Assay Details
Target Gene Details
Entrez Gene ID: | 7187 |
Gene Name: | TNF receptor associated factor 3 |
Gene Aliases: |
CAP-1, CAP1, CD40bp, CRAF1, IIAE5, LAP1 |
Location: |
Chr.14:102777479-102911500 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRAF3 | NM_001199427.1 | NP_001186356.1 | ||
| NM_003300.3 | NP_003291.2 | |||
| NM_145725.2 | NP_663777.1 | |||
| NM_145726.2 | NP_663778.1 | |||
| XM_011537116.2 | XP_011535418.1 | |||
| XM_011537117.2 | XP_011535419.1 | |||
| XM_011537118.2 | XP_011535420.1 | |||
| XM_017021620.1 | XP_016877109.1 | |||
| AK303172.1 | ||||
| BX247977.1 | CAD62311.1 | |||
| U15637.1 | AAA56753.1 | |||
| U21092.1 | AAC50112.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv1008810 | Chr.14:102784744 - 102795643 on Build GRCh38 | Deletion |
|
| esv3635571 | Chr.14:102772846 - 102911700 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map