Assay Details
Target Gene Details
Entrez Gene ID: | 55449 |
Gene Name: | DHRS4 antisense RNA 1 |
Gene Aliases: |
AS1DHRS4, C14orf167, DHRS4AS1, PRO1488 |
Location: |
Chr.14:23938731-23955112 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHRS4-AS1 | NR_023921.2 | 2 | 1591 | |
| NR_023922.2 | 2 | 1332 | ||
| NR_023923.2 | 3 | 1300 | ||
| NR_023924.2 | 2 | 1391 | ||
| BX247982.1 | 2 | 1061 | CAD62316.1 | |
| BX647964.1 | 3 | 1270 | ||
| HQ845197.1 | 3 | 1286 | ||
| HQ845198.1 | 3 | 1286 | ||
| HQ845199.1 | 2 | 1371 | ||
| JQ979294.1 | 3 | 1286 | ||
| KJ545484.1 | 2 | 1227 | ||
| KJ574270.1 | 2 | 1018 | ||
| KJ574271.1 | 2 | 1151 | ||
| KJ574272.1 | 2 | 1150 | ||
| KJ574273.1 | 2 | 1310 | ||
| KJ574274.1 | 2 | 1354 | ||
| KJ599455.1 | 2 | 1084 | ||
| KJ599456.1 | 2 | 1341 | ||
| KJ599457.1 | 2 | 1043 | ||
| KJ599458.1 | 1 | 837 | ||
| KJ599459.1 | 3 | 1259 | ||
| KJ599460.1 | 2 | 1385 | ||
| KJ599461.1 | 3 | 1269 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1217 | Chr.14:23931356 - 24043480 on Build GRCh38 | Deletion |
|
| nsv89 | Chr.14:23935989 - 24006004 on Build GRCh38 | Deletion |
|

Genomic Map