Assay Details
Target Gene Details
Entrez Gene ID: | 5527 |
Gene Name: | protein phosphatase 2 regulatory subunit B'gamma |
Gene Aliases: |
B56G, PR61G |
Location: |
Chr.14:101760573-101927992 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R5C | NM_001161725.1 | NP_001155197.1 | ||
| NM_001161726.1 | NP_001155198.1 | |||
| XM_005267819.1 | XP_005267876.1 | |||
| XM_005267820.1 | XP_005267877.1 | |||
| XM_005267822.3 | XP_005267879.1 | |||
| XM_005267823.1 | XP_005267880.1 | |||
| XM_005267824.1 | XP_005267881.1 | |||
| XM_005267827.1 | XP_005267884.1 | |||
| AI692258.1 | ||||
| AK131391.1 | BAD18542.1 | |||
| AK302470.1 | ||||
| AL834350.1 |
More Information
Set Membership: |
|

Genomic Map