Assay Details
Target Gene Details
Entrez Gene ID: | 341880 |
Gene Name: | solute carrier family 35 member F4 |
Gene Aliases: |
C14orf36, c14_5373 |
Location: |
Chr.14:57563922-57982329 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35F4 | NM_001206920.1 | NP_001193849.1 | ||
| NM_001306087.1 | NP_001293016.1 | |||
| XM_011536720.2 | XP_011535022.1 | |||
| XM_011536721.2 | XP_011535023.1 | |||
| XM_011536723.2 | XP_011535025.1 | |||
| XM_011536724.2 | XP_011535026.1 | |||
| XM_011536725.1 | XP_011535027.1 | |||
| XM_017021258.1 | XP_016876747.1 | |||
| XM_017021259.1 | XP_016876748.1 | |||
| XM_017021260.1 | XP_016876749.1 | |||
| XM_017021261.1 | XP_016876750.1 | |||
| BC101318.1 | ||||
| BC101320.1 | ||||
| BC101321.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046422 | Chr.14:57198718 - 58129812 on Build GRCh38 | Gain |
|
| nsv1043109 | Chr.14:57545677 - 57618153 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map