Assay Details
Target Gene Details
Entrez Gene ID: | 5527 |
Gene Name: | protein phosphatase 2 regulatory subunit B'gamma |
Gene Aliases: |
B56G, PR61G |
Location: |
Chr.14:101760573-101927992 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R5C | XM_005267819.1 | XP_005267876.1 | ||
| XM_005267822.3 | XP_005267879.1 | |||
| XM_005267823.1 | XP_005267880.1 | |||
| XM_005267824.1 | XP_005267881.1 | |||
| XM_005267827.1 | XP_005267884.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1070658 | Chr.14:101760362 - 101762063 on Build GRCh38 | Deletion |
|
| nsv952250 | Chr.14:101760264 - 101762063 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map