Assay Details
Target Gene Details
Entrez Gene ID: | 55449 |
Gene Name: | DHRS4 antisense RNA 1 |
Gene Aliases: |
AS1DHRS4, C14orf167, DHRS4AS1, PRO1488 |
Location: |
Chr.14:23938731-23955112 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHRS4-AS1 | NR_023921.2 | |||
| NR_023922.2 | ||||
| NR_023923.2 | ||||
| NR_023924.2 | ||||
| BF206895.1 | ||||
| BX247982.1 | CAD62316.1 | |||
| BX447332.2 | ||||
| BX482101.1 | ||||
| BX647964.1 | ||||
| DA439818.1 | ||||
| DB254318.1 | ||||
| HQ845197.1 | ||||
| HQ845198.1 | ||||
| HQ845199.1 | ||||
| JQ979294.1 | ||||
| KJ545484.1 | ||||
| KJ574270.1 | ||||
| KJ574271.1 | ||||
| KJ574272.1 | ||||
| KJ574273.1 | ||||
| KJ574274.1 | ||||
| KJ599455.1 | ||||
| KJ599456.1 | ||||
| KJ599457.1 | ||||
| KJ599459.1 | ||||
| KJ599460.1 | ||||
| KJ599461.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1217 | Chr.14:23931356 - 24043480 on Build GRCh38 | Deletion |
|
| nsv89 | Chr.14:23935989 - 24006004 on Build GRCh38 | Deletion |
|

Genomic Map