Assay Details
Target Gene Details
Entrez Gene ID: | 9154 |
Gene Name: | solute carrier family 28 member 1 |
Gene Aliases: |
CNT1, HCNT1 |
Location: |
Chr.15:84884351-84975649 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524050 | Chr.15:84864333 - 84935498 on Build GRCh38 | Loss |
|
| nsv471257 | Chr.15:84869224 - 84919085 on Build GRCh38 | Gain |
|
| esv2672879 | Chr.15:84183834 - 85188520 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map