Assay Details
Target Gene Details
Entrez Gene ID: | 197135 |
Gene Name: | PAT1 homolog 2 |
Gene Aliases: |
Pat1a, hPat1a |
Location: |
Chr.15:44665732-44711389 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PATL2 | XM_011521338.2 | 1 | 308 | XP_011519640.1 |
| XM_011521339.2 | 1 | 308 | XP_011519641.1 | |
| XM_011521340.2 | 1 | 308 | XP_011519642.1 | |
| XM_011521343.2 | 1 | 308 | XP_011519645.1 | |
| XM_011521345.2 | 1 | 308 | XP_011519647.1 | |
| AK130783.1 | 1 | 235 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832990 | Chr.15:44698567 - 44870516 on Build GRCh38 | Gain |
|
| esv2760029 | Chr.15:44676670 - 45130644 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map