Assay Details
Target Gene Details
Entrez Gene ID: | 283673 |
Gene Name: | Ewing sarcoma associated transcript 1 |
Gene Aliases: |
LINC00277, NCRNA00277, TMEM84 |
Location: |
Chr.15:69080850-69095824 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EWSAT1 | NR_026949.1 | |||
| AK091087.1 | BAC03580.1 | |||
| AK097560.1 | ||||
| AK292174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1050604 | Chr.15:69065909 - 69107327 on Build GRCh38 | Gain |
|
| nsv569808 | Chr.15:69035886 - 69100053 on Build GRCh38 | Loss |
|

Genomic Map