Assay Details
Target Gene Details
Entrez Gene ID: | 567 |
Gene Name: | beta-2-microglobulin |
Gene Aliases: |
IMD43 |
Location: |
Chr.15:44711487-44718159 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B2M | NM_004048.2 | NP_004039.1 | ||
| XM_005254549.3 | XP_005254606.1 | |||
| AB021288.1 | BAA35182.1 | |||
| AK303560.1 | ||||
| AK315776.1 | ||||
| AY007153.1 | AAG02006.1 | |||
| AY187687.1 | AAO20842.1 | |||
| BC032589.1 | AAH32589.1 | |||
| BC064910.1 | AAH64910.1 | |||
| BU658737.1 | ||||
| CR457066.1 | CAG33347.1 | |||
| DQ839493.1 | ||||
| DQ839494.1 | ||||
| DQ884406.1 | ||||
| S54761.1 | AAB25312.1 | |||
| S82297.1 | AAD14387.1 | |||
| S82300.1 | AAD14388.1 | |||
| V00567.1 | CAA23830.1 | |||
| X07621.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832990 | Chr.15:44698567 - 44870516 on Build GRCh38 | Gain |
|
| esv2760029 | Chr.15:44676670 - 45130644 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map