Assay Details
Target Gene Details
Entrez Gene ID: | 54893 |
Gene Name: | myotubularin related protein 10 |
Gene Aliases: |
- |
Location: |
Chr.15:30919764-30991604 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTMR10 | NM_017762.2 | NP_060232.2 | ||
XM_005254508.2 | XP_005254565.1 | |||
XM_011521737.2 | XP_011520039.1 | |||
XM_017022384.1 | XP_016877873.1 | |||
AK296926.1 | ||||
AK303672.1 | ||||
AL832137.1 | ||||
AL832603.1 | ||||
KJ534882.1 | ||||
KJ534883.1 | ||||
KJ534987.1 | ||||
KJ535012.1 | ||||
KJ535030.1 | ||||
KJ535071.1 | ||||
KJ535075.1 | ||||
KJ535083.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832951 | Chr.15:30839036 - 31012266 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv525795 | Chr.15:30644082 - 31032328 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3569376 | Chr.15:30781533 - 30987140 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv33507 | Chr.15:30891121 - 31178820 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |
nsv521842 | Chr.15:30848497 - 30980467 on Build GRCh38 | Gain |
![]() ![]() |
nsv1046874 | Chr.15:30715930 - 31002451 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv3636036 | Chr.15:30818302 - 31038636 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv832952 | Chr.15:30912223 - 31041795 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
