Assay Details
Target Gene Details
Entrez Gene ID: | 3418 |
Gene Name: | isocitrate dehydrogenase (NADP(+)) 2, mitochondrial |
Gene Aliases: |
D2HGA2, ICD-M, IDH, IDHM, IDP, IDPM, mNADP-IDH |
Location: |
Chr.15:90083978-90102554 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDH2 | NM_001289910.1 | NP_001276839.1 | ||
| NM_001290114.1 | NP_001277043.1 | |||
| NM_002168.3 | NP_002159.2 | |||
| AK222916.1 | BAD96636.1 | |||
| AK294148.1 | ||||
| AK299987.1 | ||||
| AK312627.1 | ||||
| AK316388.1 | ||||
| BC009244.2 | AAH09244.1 | |||
| BC071828.1 | AAH71828.1 | |||
| X69433.1 | CAA49208.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3637213 | Chr.15:90084539 - 90105288 on Build GRCh38 | Gain |
|
| dgv112e55 | Chr.15:90079272 - 90249274 on Build GRCh38 | Gain |
|
| esv2760054 | Chr.15:90022724 - 90266401 on Build GRCh38 | Gain |
|
| esv3569864 | Chr.15:90056527 - 90089206 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115464181] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map