Assay Details
Target Gene Details
Entrez Gene ID: | 101926928 |
Gene Name: | cervical cancer-associated transcript 37 |
Gene Aliases: |
- |
Location: |
Chr.15:91463339-91494850 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRAT37 | NR_110106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833095 | Chr.15:91463563 - 91607946 on Build GRCh38 | Loss |
|
| nsv833094 | Chr.15:91288643 - 91497082 on Build GRCh38 | Gain |
|
| esv2760055 | Chr.15:91288620 - 91497082 on Build GRCh38 | Gain |
|

Genomic Map