Assay Details
Target Gene Details
Entrez Gene ID: | 84465 |
Gene Name: | multiple EGF like domains 11 |
Gene Aliases: |
- |
Location: |
Chr.15:65895296-66253737 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MEGF11 | NM_032445.2 | NP_115821.2 | ||
| XM_017022670.1 | XP_016878159.1 | |||
| XM_017022671.1 | XP_016878160.1 | |||
| XM_017022672.1 | XP_016878161.1 | |||
| XM_017022673.1 | XP_016878162.1 | |||
| AB058677.1 | ||||
| AB300051.1 | ||||
| BC117419.1 | ||||
| BC126313.1 |
Target Gene Details
Entrez Gene ID: | 100422905 |
Gene Name: | microRNA 4311 |
Gene Aliases: |
- |
Location: |
Chr.15:66040233-66040332 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR4311 | NR_036196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509572 | Chr.15:65970332 - 66112995 on Build GRCh38 | Insertion |
|
| nsv1047176 | Chr.15:66036143 - 66310781 on Build GRCh38 | Gain |
|
| nsv1572 | Chr.15:66012939 - 66058057 on Build GRCh38 | Deletion |
|
| esv2760035 | Chr.15:65888447 - 66058047 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map