Assay Details
Target Gene Details
Entrez Gene ID: | 83886 |
Gene Name: | protease, serine 27 |
Gene Aliases: |
CAPH2, MPN |
Location: |
Chr.16:2712418-2720551 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS27 | NM_001318395.1 | NP_001305324.1 | ||
| NM_031948.4 | NP_114154.1 | |||
| AB056161.1 | BAB85497.1 | |||
| AJ306593.1 | CAC35467.1 | |||
| AK310214.1 | ||||
| AY030095.1 | AAK38168.1 | |||
| AY359106.1 | AAQ89464.1 | |||
| BC034294.2 | AAH34294.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
|
| esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss |
|
| nsv471073 | Chr.16:2687070 - 2773569 on Build GRCh38 | Loss |
|
| nsv833123 | Chr.16:2561247 - 2764988 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74375707] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map