Assay Details
Target Gene Details
Entrez Gene ID: | 146227 |
Gene Name: | brain expressed, associated with NEDD4, 1 |
Gene Aliases: |
BEAN, SCA31 |
Location: |
Chr.16:66427297-66495288 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BEAN1 | NM_001136106.4 | NP_001129578.1 | ||
| NM_001178020.2 | NP_001171491.1 | |||
| NM_001197224.3 | NP_001184153.1 | |||
| NM_001197225.3 | NP_001184154.1 | |||
| XM_011522883.1 | XP_011521185.1 | |||
| XM_011522884.1 | XP_011521186.1 | |||
| XM_011522887.2 | XP_011521189.1 | |||
| XM_011522888.1 | XP_011521190.1 | |||
| XM_011522889.1 | XP_011521191.1 | |||
| XM_011522890.1 | XP_011521192.1 | |||
| XM_011522891.1 | XP_011521193.1 | |||
| XM_011522894.2 | XP_011521196.1 | |||
| XM_011522895.1 | XP_011521197.1 | |||
| AB472390.1 | ||||
| AB472391.1 | ||||
| AB472392.1 | ||||
| AB472395.1 | ||||
| AB472396.1 | ||||
| AK056142.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833264 | Chr.16:66298692 - 66471069 on Build GRCh38 | Gain |
|
| nsv1838 | Chr.16:66420117 - 66453805 on Build GRCh38 | Insertion |
|

Genomic Map