Assay Details
Target Gene Details
Entrez Gene ID: | 102467146 |
Gene Name: | MAF transcriptional regulator RNA |
Gene Aliases: |
linc-MAF-4, lincMAF4 |
Location: |
Chr.16:79721312-79770532 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAFTRR | NR_104663.1 | |||
| BX108785.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3639248 | Chr.16:79709018 - 79770282 on Build GRCh38 | Loss |
|
| esv3572208 | Chr.16:79668676 - 80101641 on Build GRCh38 | Gain |
|
| nsv833296 | Chr.16:79658339 - 79855830 on Build GRCh38 | Loss |
|
| nsv833297 | Chr.16:79698604 - 79886522 on Build GRCh38 | Gain |
|
| nsv573285 | Chr.16:79124285 - 80093429 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79035328] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map