Assay Details
Target Gene Details
Entrez Gene ID: | 2734 |
Gene Name: | golgi glycoprotein 1 |
Gene Aliases: |
CFR-1, ESL-1, MG-160, MG160 |
Location: |
Chr.16:74447427-74607144 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLG1 | NM_001145666.1 | NP_001139138.1 | ||
| NM_001145667.1 | NP_001139139.1 | |||
| NM_012201.5 | NP_036333.2 | |||
| NR_027264.1 | ||||
| NR_027265.1 | ||||
| AK025457.1 | ||||
| AK026972.1 | ||||
| AK027032.1 | ||||
| AK304156.1 | ||||
| BC060822.1 | AAH60822.1 | |||
| U28811.1 | AAB02178.1 | |||
| U64791.1 | AAB06460.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833279 | Chr.16:74456848 - 74629175 on Build GRCh38 | Loss |
|
| nsv833278 | Chr.16:74382046 - 74550225 on Build GRCh38 | Loss |
|
| nsv1065964 | Chr.16:74054192 - 74541706 on Build GRCh38 | Gain |
|
| esv3639067 | Chr.16:74445112 - 74488116 on Build GRCh38 | Gain |
|
| esv2758656 | Chr.16:74300882 - 74553287 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map