Assay Details
Target Gene Details
Entrez Gene ID: | 27324 |
Gene Name: | TOX high mobility group box family member 3 |
Gene Aliases: |
CAGF9, TNRC9 |
Location: |
Chr.16:52436415-52547802 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOX3 | NM_001080430.3 | 7 | 2204 | NP_001073899.2 |
| NM_001146188.2 | 8 | 2206 | NP_001139660.1 | |
| XM_005255892.3 | 7 | 2227 | XP_005255949.1 | |
| XM_011523002.2 | 7 | 2965 | XP_011521304.1 | |
| XM_017023142.1 | 7 | 2280 | XP_016878631.1 | |
| AK025084.1 | 1 | 924 | ||
| AK027006.1 | 6 | 1939 | ||
| AK095095.1 | 2 | 2499 | ||
| AK299202.1 | 8 | 2206 | ||
| U80736.1 | 3 | 1298 | AAB91435.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066719 | Chr.16:52261069 - 53107993 on Build GRCh38 | Loss |
|
| nsv833229 | Chr.16:52331311 - 52488445 on Build GRCh38 | Loss |
|
| nsv457488 | Chr.16:52434324 - 52488805 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs144919658] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map