Assay Details
Target Gene Details
| Entrez Gene ID: | 23300 | 
| Gene Name: | ATM interactor | 
| Gene Aliases: | ASCIZ, ZNF822 | 
| Location: | Chr.16:81035853-81047346 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| ATMIN | NM_015251.2 | NP_056066.2 | ||
| AK290943.1 | ||||
| BC002701.2 | ||||
| BX343387.2 | ||||
| BX375235.2 | ||||
| CF593326.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv573338 | Chr.16:80986788 - 81073339 on Build GRCh38 | Loss |  C16orf46  ATMIN  CMC2  CENPN | 
| nsv1897 | Chr.16:81026961 - 81071655 on Build GRCh38 | Deletion |  C16orf46  ATMIN  CENPN | 
| esv3639306 | Chr.16:80981176 - 81044463 on Build GRCh38 | Gain |  ATMIN  CMC2  CENPN | 
| nsv1062724 | Chr.16:81018707 - 81103402 on Build GRCh38 | Gain |  C16orf46  ATMIN  GCSH  PKD1L2  CENPN | 
| esv2422360 | Chr.16:81032428 - 81147122 on Build GRCh38 | Duplication |  C16orf46  ATMIN  GCSH  PKD1L2  CENPN | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top