Assay Details
Target Gene Details
Entrez Gene ID: | 10052 |
Gene Name: | gap junction protein gamma 1 |
Gene Aliases: |
CX45, GJA7 |
Location: |
Chr.17:44794988-44830811 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJC1 | NM_001080383.1 | NP_001073852.1 | ||
| NM_005497.3 | NP_005488.2 | |||
| XM_005256920.1 | XP_005256977.1 | |||
| CN299428.1 | ||||
| DA845933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3554461 | Chr.17:44811199 - 44814402 on Build GRCh38 | Deletion |
|
| esv2674378 | Chr.17:44811315 - 44814425 on Build GRCh38 | Deletion |
|
| nsv833456 | Chr.17:44639788 - 44825873 on Build GRCh38 | Gain+Loss |
|
| nsv828021 | Chr.17:44786920 - 44829084 on Build GRCh38 | Gain |
|
| esv3640655 | Chr.17:44810918 - 44814459 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map