Assay Details
Target Gene Details
Entrez Gene ID: | 124540 |
Gene Name: | musashi RNA binding protein 2 |
Gene Aliases: |
MSI2H |
Location: |
Chr.17:57255851-57684689 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSI2 | NM_001322250.1 | NP_001309179.1 | ||
| NM_001322251.1 | NP_001309180.1 | |||
| NM_138962.3 | NP_620412.1 | |||
| NM_170721.1 | NP_733839.1 | |||
| XM_005257014.3 | XP_005257071.1 | |||
| XM_005257015.3 | XP_005257072.1 | |||
| XM_011524286.1 | XP_011522588.1 | |||
| XM_017024147.1 | XP_016879636.1 | |||
| XM_017024148.1 | XP_016879637.1 | |||
| AK093888.1 | BAC04244.1 | |||
| AK295060.1 | ||||
| BC001526.2 | AAH01526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1133410 | Chr.17:57256139 - 57257339 on Build GRCh38 | Deletion |
|
| nsv2106 | Chr.17:57231044 - 57263652 on Build GRCh38 | Insertion |
|
| esv33941 | Chr.17:57012706 - 57716106 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map