Assay Details
Target Gene Details
Entrez Gene ID: | 1468 |
Gene Name: | solute carrier family 25 member 10 |
Gene Aliases: |
DIC |
Location: |
Chr.17:81712236-81721016 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A10 | NM_001270888.1 | NP_001257817.1 | ||
| NM_001270953.1 | NP_001257882.1 | |||
| NM_012140.4 | NP_036272.2 | |||
| XM_017024220.1 | XP_016879709.1 | |||
| AI081980.1 | ||||
| AJ131613.1 | CAB59892.1 | |||
| AK075249.1 | BAC11497.1 | |||
| AK303808.1 | ||||
| BC007355.2 | AAH07355.1 | |||
| BC015797.1 | AAH15797.1 | |||
| BM561945.1 | ||||
| KU177998.1 | ||||
| KU177999.1 | ||||
| KU178000.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26223 | Chr.17:80659041 - 81736487 on Build GRCh38 | Gain+Loss |
|
| nsv428351 | Chr.17:81152631 - 83064073 on Build GRCh38 | Gain |
|
| nsv470620 | Chr.17:81516973 - 82064485 on Build GRCh38 | Loss |
|
| nsv952390 | Chr.17:81637175 - 81860124 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73354156] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map