Assay Details
Target Gene Details
Entrez Gene ID: | 2905 |
Gene Name: | glutamate ionotropic receptor NMDA type subunit 2C |
Gene Aliases: |
GluN2C, NMDAR2C, NR2C |
Location: |
Chr.17:74842021-74860843 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIN2C | NM_000835.4 | NP_000826.2 | ||
| NR_103735.1 | ||||
| XM_006721845.3 | XP_006721908.1 | |||
| XM_006721846.3 | XP_006721909.1 | |||
| XM_011524686.2 | XP_011522988.1 | |||
| XM_011524687.2 | XP_011522989.1 | |||
| XM_011524688.2 | XP_011522990.1 | |||
| XM_011524689.2 | XP_011522991.1 | |||
| XM_017024544.1 | XP_016880033.1 | |||
| AB208799.1 | BAD92036.1 | |||
| BC140801.1 | ||||
| L76224.1 | AAA88096.1 | |||
| U77782.1 | AAB71624.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833540 | Chr.17:74838971 - 75019365 on Build GRCh38 | Loss |
|
| nsv518292 | Chr.17:74810882 - 74982818 on Build GRCh38 | Loss |
|
| nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion |
|
| nsv519902 | Chr.17:74840873 - 74854994 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map