Assay Details
Target Gene Details
Entrez Gene ID: | 63897 |
Gene Name: | HEAT repeat containing 6 |
Gene Aliases: |
ABC1 |
Location: |
Chr.17:60041363-60078931 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEATR6 | XM_005257570.3 | 21 | 5802 | XP_005257627.1 |
| XM_011525110.2 | 19 | 5501 | XP_011523412.1 | |
| XM_011525111.2 | 19 | 5463 | XP_011523413.1 | |
| XM_017024936.1 | 20 | 5875 | XP_016880425.1 | |
| XM_017024937.1 | 20 | 7551 | XP_016880426.1 | |
| XM_017024938.1 | 20 | 5607 | XP_016880427.1 | |
| XM_017024939.1 | 20 | 5878 | XP_016880428.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758700 | Chr.17:59918286 - 60081801 on Build GRCh38 | Gain+Loss |
|
| nsv833502 | Chr.17:59895972 - 60081795 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map