Assay Details
Target Gene Details
Entrez Gene ID: | 27092 |
Gene Name: | calcium voltage-gated channel auxiliary subunit gamma 4 |
Gene Aliases: |
- |
Location: |
Chr.17:66964864-67033402 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNG4 | NM_014405.3 | NP_055220.1 | ||
| AF162692.1 | AAF14538.1 | |||
| AK315149.1 | ||||
| BC034532.1 | AAH34532.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469731 | Chr.17:66966354 - 67152566 on Build GRCh38 | Loss |
|
| nsv833521 | Chr.17:66858900 - 67027619 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map