Assay Details
Target Gene Details
Entrez Gene ID: | 124512 |
Gene Name: | methyltransferase like 23 |
Gene Aliases: |
C17orf95, MRT44 |
Location: |
Chr.17:76725874-76733881 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833550 | Chr.17:76712038 - 76886290 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map