Assay Details
Target Gene Details
Entrez Gene ID: | 9121 |
Gene Name: | solute carrier family 16 member 5 |
Gene Aliases: |
MCT5, MCT6 |
Location: |
Chr.17:75087727-75106590 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC16A5 | NM_001271765.1 | NP_001258694.1 | ||
| NM_004695.3 | NP_004686.1 | |||
| XM_005257790.1 | XP_005257847.1 | |||
| XM_011525462.1 | XP_011523764.1 | |||
| XM_017025293.1 | 6 | 2501 | XP_016880782.1 | |
| AK000416.1 | ||||
| AK092512.1 | ||||
| AW452009.1 | ||||
| BC009684.1 | AAH09684.1 | |||
| BC033611.1 | AAH33611.1 | |||
| U59299.1 | AAC52013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470602 | Chr.17:75090871 - 75208042 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map