Assay Details
Target Gene Details
Entrez Gene ID: | 8526 |
Gene Name: | diacylglycerol kinase epsilon |
Gene Aliases: |
AHUS7, DAGK5, DAGK6, DGK, NPHS7 |
Location: |
Chr.17:56834099-56869565 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DGKE | NM_003647.2 | NP_003638.1 | ||
| XM_011525394.2 | XP_011523696.1 | |||
| XM_011525395.2 | XP_011523697.1 | |||
| XM_011525396.2 | XP_011523698.1 | |||
| XM_017025243.1 | XP_016880732.1 | |||
| XM_017025244.1 | XP_016880733.1 | |||
| XM_017025245.1 | XP_016880734.1 | |||
| AK293039.1 | ||||
| AK310668.1 | ||||
| AL555194.3 | ||||
| BC022297.1 | AAH22297.1 | |||
| U49379.1 | AAC50497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32884 | Chr.17:56683534 - 57002132 on Build GRCh38 | Loss |
|
| nsv833495 | Chr.17:56675225 - 56862382 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map