Assay Details
Target Gene Details
| Entrez Gene ID: | 79922 | 
| Gene Name: | mitochondrial rRNA methyltransferase 1 | 
| Gene Aliases: | - | 
| Location: | Chr.17:36601581-36634698 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 6 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| MRM1 | XM_011525275.2 | XP_011523577.1 | ||
| XM_011525276.2 | XP_011523578.1 | |||
| XM_017025112.1 | XP_016880601.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| nsv833429 | Chr.17:36605619 - 36790120 on Build GRCh38 | Gain |  MRM1 | 
| dgv3156n100 | Chr.17:36589879 - 36646211 on Build GRCh38 | Gain |  MRM1  DHRS11 | 
| esv3640474 | Chr.17:36564700 - 36908342 on Build GRCh38 | Gain |  LOC102723471  MRM1  DHRS11  GGNBP2 | 
| esv2758686 | Chr.17:36588163 - 36790120 on Build GRCh38 | Gain+Loss |  MRM1  DHRS11  GGNBP2 | 
| esv3640475 | Chr.17:36602406 - 36633068 on Build GRCh38 | Gain |  MRM1 | 
| nsv469760 | Chr.17:36519312 - 36683773 on Build GRCh38 | Loss |  MYO19  MRM1  DHRS11  PIGW  GGNBP2 | 
 Back To Top
 Back To TopMore Information
Additional Information:
				    	 
					    	
					        	
					            	For this assay, SNP(s) [rs75008450] are located under a primer or probe  sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
					        	
					    	            
				    	
			    	
			    
			| Set Membership: |  Intragenic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top