Assay Details
Target Gene Details
Entrez Gene ID: | 390858 |
Gene Name: | O-acyltransferase like, pseudogene |
Gene Aliases: |
ACYL3P |
Location: |
Chr.18:59035679-59053214 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
OACYLP | NR_024021.2 | |||
DV080257.1 | ||||
DV080258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv130933 | Chr.18:59029174 - 59037657 on Build GRCh38 | Deletion |
![]() |
nsv833664 | Chr.18:58947109 - 59093282 on Build GRCh38 | Loss |
![]() ![]() |
