Assay Details
Target Gene Details
Entrez Gene ID: | 65258 |
Gene Name: | metallophosphoesterase 1 |
Gene Aliases: |
PGAP5 |
Location: |
Chr.18:11883471-11908797 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758713 | Chr.18:11897024 - 12076103 on Build GRCh38 | Loss |
|
| nsv1057995 | Chr.18:10626172 - 11923368 on Build GRCh38 | Gain |
|
| nsv1059775 | Chr.18:11892846 - 11911144 on Build GRCh38 | Loss |
|
| nsv833590 | Chr.18:11757330 - 11924540 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115556565] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map