Assay Details
Target Gene Details
Entrez Gene ID: | 64762 |
Gene Name: | GRB2 associated regulator of MAPK1 subtype 1 |
Gene Aliases: |
C18orf11, FAM59A, GAREM, Gm944 |
Location: |
Chr.18:32263521-32470518 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GAREM1 | NM_001242409.1 | NP_001229338.1 | ||
NM_022751.2 | NP_073588.1 | |||
XM_017025919.1 | XP_016881408.1 | |||
AK025263.1 | BAB15094.1 | |||
AK025660.1 | ||||
AK297430.1 | ||||
AL834491.1 | CAD39149.1 | |||
BC005074.1 | AAH05074.1 | |||
BC121067.1 | ||||
BC121068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1056007 | Chr.18:32132011 - 32319601 on Build GRCh38 | Gain |
![]() ![]() |
nsv833616 | Chr.18:32212373 - 32384249 on Build GRCh38 | Gain |
![]() ![]() |
nsv528846 | Chr.18:32252184 - 32310079 on Build GRCh38 | Gain |
![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
