Assay Details
Target Gene Details
Entrez Gene ID: | 9229 |
Gene Name: | DLG associated protein 1 |
Gene Aliases: |
DAP-1, DAP-1-ALPHA, DAP-1-BETA, DAP1, DLGAP1A, DLGAP1B, GKAP, SAPAP1 |
Location: |
Chr.18:3496032-4455307 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLGAP1 | NM_001242761.1 | NP_001229690.1 | ||
NM_004746.3 | NP_004737.2 | |||
XM_005258171.2 | XP_005258228.1 | |||
XM_005258172.1 | XP_005258229.1 | |||
XM_011525770.2 | XP_011524072.1 | |||
XM_011525771.2 | XP_011524073.1 | |||
AK304101.1 | ||||
AK316099.1 |
Target Gene Details
Entrez Gene ID: | 201477 |
Gene Name: | DLGAP1 antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.18:3878180-3897069 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DLGAP1-AS3 | NR_038895.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv134e55 | Chr.18:3793886 - 4106164 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv833582 | Chr.18:3762990 - 3931071 on Build GRCh38 | Loss |
![]() ![]() ![]() |
dgv184n21 | Chr.18:3526777 - 5930980 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv2191 | Chr.18:3876907 - 3919812 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
esv2758710 | Chr.18:3677422 - 4170252 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
