Assay Details
Target Gene Details
Entrez Gene ID: | 80816 |
Gene Name: | additional sex combs like 3, transcriptional regulator |
Gene Aliases: |
BRPS, KIAA1713 |
Location: |
Chr.18:33578577-33751195 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASXL3 | NM_030632.2 | NP_085135.1 | ||
| XM_005258356.1 | XP_005258413.1 | |||
| XM_011526209.1 | XP_011524511.1 | |||
| AK131454.1 | BAD18599.1 | |||
| BE145544.1 | ||||
| DB061724.1 | ||||
| DB090653.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762001 | Chr.18:33537508 - 33709933 on Build GRCh38 | Gain |
|
| esv2668525 | Chr.18:33248704 - 33887857 on Build GRCh38 | Deletion |
|
| nsv833618 | Chr.18:33417973 - 33593061 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140455600] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map