Assay Details
Target Gene Details
Entrez Gene ID: | 9388 |
Gene Name: | lipase G, endothelial type |
Gene Aliases: |
EDL, EL, PRO719 |
Location: |
Chr.18:49560699-49592908 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIPG | NM_001308006.1 | NP_001294935.1 | ||
| NM_006033.3 | NP_006024.1 | |||
| XM_005258390.1 | XP_005258447.1 | |||
| XM_011526265.2 | XP_011524567.1 | |||
| XM_011526267.1 | XP_011524569.1 | |||
| XM_017026095.1 | XP_016881584.1 | |||
| AF118767.1 | AAD30434.1 | |||
| AK291799.1 | ||||
| AK300333.1 | ||||
| AK315252.1 | ||||
| BC060825.1 | AAH60825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv438288 | Chr.18:49575632 - 49591539 on Build GRCh38 | Loss |
|
| nsv576863 | Chr.18:49569117 - 49938639 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116032790,rs145200514] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map