Assay Details
Target Gene Details
Entrez Gene ID: | 9306 |
Gene Name: | suppressor of cytokine signaling 6 |
Gene Aliases: |
CIS-4, CIS4, HSPC060, SOCS-4, SOCS-6, SOCS4, SSI4, STAI4, STATI4 |
Location: |
Chr.18:70288901-70330199 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SOCS6 | NM_004232.3 | NP_004223.2 | ||
XM_005266783.3 | XP_005266840.1 | |||
XM_017026087.1 | XP_016881576.1 | |||
BC020082.1 | AAH20082.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv32736 | Chr.18:69135149 - 71678611 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv34014 | Chr.18:70310065 - 70807720 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
