Assay Details
Target Gene Details
Entrez Gene ID: | 171586 |
Gene Name: | abhydrolase domain containing 3 |
Gene Aliases: |
LABH3 |
Location: |
Chr.18:21650897-21704805 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD3 | NM_001308256.1 | NP_001295185.1 | ||
| NM_138340.4 | NP_612213.2 | |||
| XM_011525847.2 | XP_011524149.1 | |||
| XM_017025571.1 | XP_016881060.1 | |||
| XM_017025572.1 | XP_016881061.1 | |||
| XM_017025573.1 | XP_016881062.1 | |||
| XM_017025574.1 | XP_016881063.1 | |||
| AF007152.1 | AAC19155.1 | |||
| AK024880.1 | ||||
| AK298728.1 | ||||
| BC021196.2 | AAH21196.1 | |||
| BI918326.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2716850 | Chr.18:21667117 - 21674310 on Build GRCh38 | Deletion |
|
| esv29974 | Chr.18:21593909 - 21668731 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map