Assay Details
Target Gene Details
Entrez Gene ID: | 147645 |
Gene Name: | V-set and immunoglobulin domain containing 10 like |
Gene Aliases: |
- |
Location: |
Chr.19:51331541-51342124 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSIG10L | NM_001163922.1 | 10 | 2764 | NP_001157394.1 |
| BC049212.1 | 7 | 1429 | AAH49212.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115816586] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map