Assay Details
Target Gene Details
Entrez Gene ID: | 402665 |
Gene Name: | IgLON family member 5 |
Gene Aliases: |
- |
Location: |
Chr.19:51311848-51330848 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGLON5 | NM_001101372.1 | NP_001094842.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1060e212 | Chr.19:51306719 - 51321097 on Build GRCh38 | Loss |
|
| nsv1132479 | Chr.19:51318846 - 51323046 on Build GRCh38 | Duplication |
|
| nsv1151102 | Chr.19:51319745 - 51323546 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map