Assay Details
Target Gene Details
Entrez Gene ID: | 342979 |
Gene Name: | paralemmin 3 |
Gene Aliases: |
- |
Location: |
Chr.19:14053367-14062137 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PALM3 | NM_001145028.1 | NP_001138500.1 | ||
| XM_005259891.4 | XP_005259948.1 | |||
| XM_011527965.2 | XP_011526267.1 | |||
| AW130597.1 | ||||
| CR747190.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833761 | Chr.19:13998659 - 14136778 on Build GRCh38 | Loss |
|
| nsv578642 | Chr.19:14028192 - 14187630 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map