Assay Details
Target Gene Details
Entrez Gene ID: | 94030 |
Gene Name: | leucine rich repeat containing 4B |
Gene Aliases: |
HSM, LRIG4 |
Location: |
Chr.19:50516892-50568208 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC4B | NM_001080457.1 | 3 | 2012 | NP_001073926.1 |
| XM_005259429.4 | 3 | 2191 | XP_005259486.1 | |
| XM_006723506.3 | 4 | 2293 | XP_006723569.1 | |
| XM_011527520.2 | 3 | 3439 | XP_011525822.1 | |
| AL137451.1 | CAB70743.2 | |||
| BC019687.3 | AAH19687.1 | |||
| BC032460.1 | ||||
| BC056207.1 | 1 | 219 | AAH56207.1 | |
| BC131897.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953604 | Chr.19:50506544 - 50580143 on Build GRCh38 | Deletion |
|
| nsv509750 | Chr.19:50443078 - 50540357 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map