Assay Details
Target Gene Details
Entrez Gene ID: | 125931 |
Gene Name: | carcinoembryonic antigen related cell adhesion molecule 20 |
Gene Aliases: |
UNQ9366 |
Location: |
Chr.19:44503838-44530100 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEACAM20 | NM_001102597.2 | NP_001096067.2 | ||
| NM_001102598.2 | NP_001096068.2 | |||
| NM_001102599.2 | NP_001096069.2 | |||
| NM_001102600.2 | NP_001096070.2 | |||
| XM_011526429.2 | XP_011524731.1 | |||
| XM_011526430.2 | XP_011524732.1 | |||
| XM_011526431.2 | XP_011524733.1 | |||
| AY358129.1 | AAQ88496.1 | |||
| EU008600.1 | ||||
| EU008601.1 | ||||
| EU008602.1 | ||||
| EU008603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061350 | Chr.19:44364917 - 44531444 on Build GRCh38 | Loss |
|

Genomic Map