Assay Details
Target Gene Details
Entrez Gene ID: | 2909 |
Gene Name: | Rho GTPase activating protein 35 |
Gene Aliases: |
GRF-1, GRLF1, P190-A, P190A, p190ARhoGAP, p190RhoGAP |
Location: |
Chr.19:46861002-47005077 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP35 | XM_011526873.2 | XP_011525175.1 | ||
| XM_017026714.1 | XP_016882203.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833850 | Chr.19:46703285 - 46867053 on Build GRCh38 | Loss |
|
| dgv3608n100 | Chr.19:46757741 - 46865807 on Build GRCh38 | Gain |
|
| esv3893224 | Chr.19:46783912 - 46911729 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs201588487] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map