Assay Details
Target Gene Details
Entrez Gene ID: | 6794 |
Gene Name: | serine/threonine kinase 11 |
Gene Aliases: |
LKB1, PJS, hLKB1 |
Location: |
Chr.19:1205799-1228435 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK11 | NM_000455.4 | NP_000446.1 | ||
| XM_005259617.2 | XP_005259674.1 | |||
| XM_011528209.1 | XP_011526511.1 | |||
| AB209553.1 | BAD92790.1 | |||
| AF035625.1 | AAC39527.1 | |||
| AF217978.1 | AAG17221.1 | |||
| AK128518.1 | 1 | 700 | ||
| AK314858.1 | ||||
| BC007981.2 | AAH07981.1 | |||
| BC019334.1 | AAH19334.1 | |||
| EF644172.1 | ||||
| KU178339.1 | ||||
| KU178340.1 | ||||
| U63333.1 | AAB05809.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058677 | Chr.19:1054061 - 1283189 on Build GRCh38 | Loss |
|
| nsv578268 | Chr.19:1221162 - 1250886 on Build GRCh38 | Loss |
|
| dgv136n111 | Chr.19:1038223 - 1329640 on Build GRCh38 | Deletion |
|
| nsv9662 | Chr.19:893394 - 1317740 on Build GRCh38 | Gain |
|
| nsv470104 | Chr.19:1170446 - 1325151 on Build GRCh38 | Loss |
|
| nsv1160555 | Chr.19:887275 - 1346084 on Build GRCh38 | Deletion |
|
| dgv6196n54 | Chr.19:1222317 - 1245701 on Build GRCh38 | Loss |
|
| nsv953941 | Chr.19:1205602 - 1322401 on Build GRCh38 | Deletion |
|
| nsv470103 | Chr.19:1086044 - 1325151 on Build GRCh38 | Gain |
|
| esv2758743 | Chr.19:951641 - 1544934 on Build GRCh38 | Gain+Loss |
|
| nsv828368 | Chr.19:1223051 - 1274100 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map