Assay Details
Target Gene Details
Entrez Gene ID: | 8605 |
Gene Name: | phospholipase A2 group IVC |
Gene Aliases: |
CPLA2-gamma |
Location: |
Chr.19:48047843-48111445 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G4C | NM_001159322.1 | NP_001152794.1 | ||
| NM_001159323.1 | NP_001152795.1 | |||
| NM_003706.2 | NP_003697.2 | |||
| XM_011527431.2 | XP_011525733.1 | |||
| XM_011527432.2 | XP_011525734.1 | |||
| XM_011527433.2 | XP_011525735.1 | |||
| XM_017027411.1 | XP_016882900.1 | |||
| XM_017027412.1 | XP_016882901.1 | |||
| XM_017027413.1 | XP_016882902.1 | |||
| XM_017027414.1 | XP_016882903.1 | |||
| AF058921.1 | AAC32823.1 | |||
| AF065214.1 | AAC78835.1 | |||
| AK022440.1 | ||||
| AK295400.1 | ||||
| AK314524.1 | ||||
| BC063416.1 | AAH63416.1 | |||
| CR456816.1 | CAG33097.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833853 | Chr.19:47982539 - 48184317 on Build GRCh38 | Loss |
|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| esv34181 | Chr.19:48009377 - 48416682 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs111793606,rs112825357] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map