Assay Details
Target Gene Details
Entrez Gene ID: | 402665 |
Gene Name: | IgLON family member 5 |
Gene Aliases: |
- |
Location: |
Chr.19:51311848-51330848 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGLON5 | NM_001101372.1 | NP_001094842.1 |
More Information
Set Membership: |
|

Genomic Map