Assay Details
Target Gene Details
Entrez Gene ID: | 140686 |
Gene Name: | WAP four-disulfide core domain 3 |
Gene Aliases: |
WAP14, dJ447F3.3 |
Location: |
Chr.20:45708860-45791908 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WFDC3 | XM_017027668.1 | XP_016883157.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2670845 | Chr.20:45744160 - 45758866 on Build GRCh38 | Deletion |
|
| nsv586131 | Chr.20:45727555 - 45778454 on Build GRCh38 | Loss |
|
| esv3645940 | Chr.20:45744194 - 45758836 on Build GRCh38 | Loss |
|
| esv2751916 | Chr.20:45722292 - 45771812 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map