Assay Details
Target Gene Details
Entrez Gene ID: | 958 |
Gene Name: | CD40 molecule |
Gene Aliases: |
Bp50, CDW40, TNFRSF5, p50 |
Location: |
Chr.20:46118242-46129745 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD40 | NM_001250.5 | NP_001241.1 | ||
| NM_001302753.1 | NP_001289682.1 | |||
| NM_001322421.1 | NP_001309350.1 | |||
| NM_001322422.1 | NP_001309351.1 | |||
| NM_152854.3 | NP_690593.1 | |||
| NR_126502.1 | ||||
| NR_136327.1 | ||||
| XM_005260619.3 | XP_005260676.1 | |||
| XM_011529109.2 | XP_011527411.1 | |||
| XM_017028135.1 | XP_016883624.1 | |||
| XM_017028136.1 | XP_016883625.1 | |||
| XM_017028137.1 | XP_016883626.1 | |||
| AB209660.1 | 5 | 2218 | BAD92897.1 | |
| AJ300189.1 | CAC29424.1 | |||
| AK222896.1 | BAD96616.1 | |||
| AY225405.1 | AAO43990.1 | |||
| BC012419.1 | AAH12419.1 | |||
| BC064518.1 | AAH64518.1 | |||
| BM761221.1 | ||||
| BT019901.1 | AAV38704.1 | |||
| BX364285.2 | ||||
| KU177966.1 | ||||
| KU177967.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057686 | Chr.20:46105657 - 46434770 on Build GRCh38 | Loss |
|
| esv2758796 | Chr.20:46064276 - 46336238 on Build GRCh38 | Loss |
|
| nsv833993 | Chr.20:46091252 - 46290015 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map