Assay Details
Target Gene Details
Entrez Gene ID: | 114044 |
Gene Name: | MCM3AP antisense RNA 1 |
Gene Aliases: |
C21orf85, MCM3AP-AS, MCM3APAS, MCM3APASB, NCRNA00031 |
Location: |
Chr.21:46229231-46251701 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM3AP-AS1 | NR_002776.4 | |||
| NR_110565.1 | ||||
| NR_110566.1 | ||||
| NR_110567.1 | ||||
| AF426263.1 | ||||
| AK001370.1 | ||||
| BC059792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519904 | Chr.21:46163592 - 46461396 on Build GRCh38 | Loss |
|
| nsv509804 | Chr.21:45879599 - 46235140 on Build GRCh38 | Insertion |
|
| nsv1144958 | Chr.21:46228886 - 46229386 on Build GRCh38 | Deletion |
|
| nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain |
|
| nsv1060978 | Chr.21:46068159 - 46413726 on Build GRCh38 | Gain |
|

Genomic Map